Methyl-3-chloro-4-methylbenzoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Methyl 3-chloro-4-methylbenzoate
M2136-25G
M2136-5G
TCIAM2136-25GEA
139
EUR
TCIAM2136-25G
TCIAM2136-5G
Methyl-3-chloro-4-methylbenzoate ≥98.0% (by GC)
Methyl-3-chloro-4-methylbenzoate
Formula:
C₉H₉ClO₂ Peso molecolare: 184,62 g/mol Punto di ebollizione: 107°C |
MDL Number:
MFCD00205141 CAS Number: 56525-63-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...