2-Chloro-5-(methylcarbamoyl)phenylboronic acid ≥97%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
2-Chloro-5-(methylcarbamoyl)benzeneboronic acid
H55618.MD
H55618.03
H55618.MDEA
127
EUR
H55618.MD
H55618.03
2-Chloro-5-(methylcarbamoyl)phenylboronic acid ≥97%
2-Chloro-5-(methylcarbamoyl)phenylboronic acid
Purity: 97%
Molecular Formula: C8H9BClNO3
Formula Weight: 213.43
MDL No.: MFCD22125158
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...