Methyl-2,2-difluoro-2-(fluorosulphonyl)acetate ≥97.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₃H₃F₃O₄S Punto di ebollizione: 118°C Flash Pt: 47°C |
MDL Number:
MFCD00144316 CAS Number: 680-15-9 UN: 2920 ADR: 8,II |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...