3-Methoxypyridazine ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
M2078-1G
M2078-5G
TCIAM2078-1GEA
62
EUR
TCIAM2078-1G
TCIAM2078-5G
3-Methoxypyridazine ≥97.0% (by GC, titration analysis)
3-Methoxypyridazine
Formula:
C₅H₆N₂O Peso molecolare: 110,1139 g/mol Punto di ebollizione: 87°C |
MDL Number:
MFCD00234098 CAS Number: 19064-65-4 |
Risultati del test di specificazione
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...