Stampa…

2-Quinoxalinecarboxylic acid ≥96.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Q0081-1G Q0081-5G
TCIAQ0081-1GEA 74 EUR
TCIAQ0081-1G TCIAQ0081-5G
2-Quinoxalinecarboxylic acid ≥96.0% (by GC, titration analysis)
2-Quinoxalinecarboxylic acid
Formula: C₉H₆N₂O₂
Peso molecolare: 174,16 g/mol
Punto di fusione: 209°C
MDL Number: MFCD00012334
CAS Number: 879-65-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3eec1d7662ebb76fd81b604f8627880cbd112a7&vid=a3eec1d7662ebb76fd81b604f8627880cbd112a7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR