Stampa…

2-(Methylthio)benzimidazole ≥99.0% (by HPLC, titration analysis)

Fornitore: TCI

M0438-25G M0438-5G
TCIAM0438-25GEA 136 EUR
TCIAM0438-25G TCIAM0438-5G
2-(Methylthio)benzimidazole ≥99.0% (by HPLC, titration analysis)
2-(Methylthio)benzimidazole
Formula: C₈H₈N₂S
MDL Number: MFCD00005594
CAS Number: 7152-24-1
EINECS: 230-494-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9723b571daeafe2a05fd66d8f88255f86c5f9445&vid=9723b571daeafe2a05fd66d8f88255f86c5f9445&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 200.0 to 207.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR