Bis(pyridine)iodonium tetrafluoroborate ≥97%
Fornitore: Thermo Scientific
|
Attenzione
|
L19385.03
L19385.MD
L19385.03EA
68.9
EUR
L19385.03
L19385.MD
Bis(pyridine)iodonium tetrafluoroborate ≥97%
Bis(pyridine)iodonium tetrafluoroborate
Purity: 97%
CAS No.: 15656-28-7
Molecular Formula: C10H10BF4IN2
Formula Weight: 371.91
Storage Temperature: 2°C to 8°C
Odour: Odourless
Beilstein Reference: 4284739
MDL No.: MFCD03703393
Formula:
C₁₀H₁₀BF₄IN₂ Peso molecolare: 371,91 g/mol Punto di fusione: ∼ 160 °C (dec.) |
MDL Number:
MFCD03703393 CAS Number: 15656-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...