Stampa…

2-Chloro-3-ethylbenzoxazolium tetrafluoroborate ≥97.0% (by total nitrogen and titration analysis)

Fornitore: TCI EUROPE
Pericolo

C0905-25G C0905-5G
TCIAC0905-25GEA 224 EUR
TCIAC0905-25G TCIAC0905-5G
2-Chloro-3-ethylbenzoxazolium tetrafluoroborate ≥97.0% (by total nitrogen and titration analysis)
2-Chloro-3-ethylbenzoxazolium tetrafluoroborate
Formula: C₉H₉BClF₄NO
Punto di fusione: 131°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00011943
CAS Number: 63212-53-3
EINECS: 263-998-9
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75fabf8d4c508120fa4c3be16e4199c032e391a7&vid=75fabf8d4c508120fa4c3be16e4199c032e391a7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR