Stampa…

(±)-trans-1,2-Cyclohexanedicarboxylic anhydride ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

C1657-25G C1657-5G
TCIAC1657-25GEA 291 EUR
TCIAC1657-25G TCIAC1657-5G
(±)-trans-1,2-Cyclohexanedicarboxylic anhydride ≥98.0% (by GC, titration analysis)
(±)-trans-1,2-Cyclohexanedicarboxylic anhydride
Formula: C₈H₁₀O₃
MDL Number: MFCD00064345
CAS Number: 14166-21-3
EINECS: 238-009-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d1d791b2cd3c34519c488d68fac09694cf75510&vid=7d1d791b2cd3c34519c488d68fac09694cf75510&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Methanolysis Method) min. 98.0 %
Melting point 144.0 to 149.0 deg-C
Specific rotation [a]20/D -1.0 to +1.0 deg (C=1, Aceton)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR