Stampa…

Di-tert-butyl pyrocarbonate ∼30% in toluene

Fornitore: TCI EUROPE
Pericolo

Synonyms: Boc Anhydride, Di-tert-butyl dicarbonate, DIBOC

D3880-100G D3880-400G
TCIAD3880-100GEA 57 EUR
TCIAD3880-100G TCIAD3880-400G
Di-tert-butyl pyrocarbonate ∼30% in toluene
Di-tert-butyl pyrocarbonate
Formula: C₁₀H₁₈O₅
Peso molecolare: 218,25 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00008805
CAS Number: 24424-99-5
EINECS: 246-240-1
UN: 1992
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c1de273b7b2c9a9a94cef2cd5d518e464395006&vid=9e6f2e051c9a5717c25cc2a796821e736f1c5d60&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Concentration (Morpholine Method) 29.0 to 33.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR