Stampa…

Methyl vanillate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: Methyl 4-hydroxy-3-methoxybenzoate

M1738-25G M1738-5G
TCIAM1738-25GEA 86 EUR
TCIAM1738-25G TCIAM1738-5G
Methyl vanillate ≥98.0% (by GC)
Methyl vanillate
Formula: C₉H₁₀O₄
Punto di ebollizione: 287°C
MDL Number: MFCD00008438
CAS Number: 3943-74-6
EINECS: 223-525-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c003d1f0907b8016d392c78824f34ce5dd4d293&vid=9c003d1f0907b8016d392c78824f34ce5dd4d293&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 66.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR