2-Formylimidazole ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
I0809-5G
I0809-1G
TCIAI0809-5GEA
83
EUR
TCIAI0809-5G
TCIAI0809-1G
2-Formylimidazole ≥98.0% (by HPLC, titration analysis)
2-Formylimidazole
Formula:
C₄H₄N₂O Punto di fusione: 202°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00003544 CAS Number: 10111-08-7 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...