Stampa…
Evaporatori rotanti analogici, Hei-VAP Value
Fornitore: Heidolph Instruments GmbH & Co.KG
560-01300-04 560-01510-03 560-01500-04 560-01500-03 560-01310-04 560-01300-03 560-01110-04 560-01110-03 560-01100-04 560-01100-03 560-01310-03 560-01510-04 560-01600-03 560-01600-04 560-01610-03 560-01610-04 560-91309-04 560-91309-03 560-91300-04 560-91300-03 560-81310-04 560-81310-03 560-01305-04 560-01305-03 560-01315-04 560-01315-03
531-0788EA 3250 EUR
531-0788 531-0700 531-0789 531-0699 531-0690 531-0697 531-0787 531-0696 531-0689 531-0695 531-0698 531-0790 531-0701 531-0791 531-0702 531-0792 531-0984 HEID560-91309-03 531-0982 HEID560-91300-03 531-0980 HEID560-81310-03 HEID560-01305-04 HEID560-01305-03 HEID560-01315-04 HEID560-01315-03
Evaporatori rotanti analogici, Hei-VAP Value
Evaporatori Evaporatori rotanti
These Hei-VAP devices, with hand lift, are ideally suited for all standard applications. Parameters can be controlled with the simple turn of a knob.
  • Large dial controls for adjustment of rotation speed and bath temperature
  • Required angle and height of the evaporating flask can adjusted individually
  • Patented EasyClip flask clamp allows the simple and safe removal of the evaporating flask due to the force of unscrewing the Easy-Clip
  • Detachable operating panel enables safe and convenient handling from outside closed fume hoods
  • Separate on/off switch for heating prevents unintentional heat-up – the button is illuminated for visual control
  • FDA-compliant, transparent and UV resistant tubing, with higher chemical resistance
210 °C safety heating bath, with handles and pour spout, will power off if temperature overshoots by 5 °C or runs dry. The bath power cable coupling complies with protection class IP 67 to prevent corrosion and short circuits.
Glassware set G1
Diagonal condenser for all standard distillations; option with the most affordable price.
Glassware set G3
Vertical condenser for all standard distillations; option with the smallest space requirements, also available as XL version (condensing surface of 2200 cm² for up to 40% higher evaporation rates).
Glassware set G5
Dry ice condenser for low boiling solvents.
Glassware set G6
Vertical condenser; centrepiece with valve for reflux distillation.
As standard, models are supplied with a detachable control panel with a 1,3 m cable. Therefore parameters can be changed outside of a closed fume hood, providing greater operator safety and ultimate process control. An optional remote control operating panel allows the user to step away from the evaporator and still monitor all process data increasing flexibility at work.
Informazioni per l'ordine: Supplied with water/oil bath, 1 L evaporation flasks (29/32), 1 L receiving flasks (35/20) and condenser glassware set.
Please note: A vacuum source, vacuum-check valve and cooling system are required but not supplied. Please contact your local Avantor sales office for advice and details.
Attenzione: * con plastica trasparente