Stampa…
Ladies combat trousers, C099
Fornitore: Portwest
C099NARL C099NARXXL C099NARXS C099NARXXXL C099NARS C099NARM C099NARXL
PRTWC099NARLEA 27 EUR
PRTWC099NARL PRTWC099NARXXL PRTWC099NARXS PRTWC099NARXXXL PRTWC099NARS PRTWC099NARM PRTWC099NARXL
Ladies combat trousers, C099
Abbigliamento da lavoro
These comfortable and stylish trousers are made of Kingsmill fabric (65% polyester/35% cotton) which guarantees high performance and maximum comfort. The material is fully pre-shrunk, has superior dye retention and all the features you would expect from a modern workwear fabric.
  • Hook and bar closure
  • Back elasticated waist for extra freedom of movement
  • Two side pockets and a combination thigh and mobile phone pocket
Weight: 210 g/m²
Visualizza i codici
Specifiche
Modello C099
Applicazioni per abiti General Purpose,Industrial
Environmentally Preferable
Risk area
Genere Per le donne
Sleeve or leg length Long
Tipo di chiusura Con Velcro®
Tessuto Kingsmill fabric (65% polyester, 35% cotton)
Peso 210 g/m²