Stampa…
General purpose gloves, A120
Fornitore: Portwest
A120GRRM A120BKRL A120WHRS A120WHRM A120WHRL A120GRRXXL A120GRRXS A120WHRXS A120GRRS A120WHRXXL A120GRRL A120BKRXXL A120BKRXS A120BKRXL A120BKRS A120BKRM A120GRRXL A120WHRXL
PRTWA120GRRMEA 15 EUR
PRTWA120GRRM PRTWA120BKRL PRTWA120WHRS PRTWA120WHRM PRTWA120WHRL PRTWA120GRRXXL PRTWA120GRRXS PRTWA120WHRXS PRTWA120GRRS PRTWA120WHRXXL PRTWA120GRRL PRTWA120BKRXXL PRTWA120BKRXS PRTWA120BKRXL PRTWA120BKRS PRTWA120BKRM PRTWA120GRRXL PRTWA120WHRXL
General purpose gloves, A120
Guanti
Seamless knitted nylon liners with polyurethane palm and finger coating. Ideal for intricate tasks where dexterity and breathability are essential.
  • Open back for extra ventilation, knitted wrist
  • High level of dexterity for the most tactile of applications
  • 13 gauge liner for a perfect fit
Typical applications include: electronics, inspection, computer assembly, precision engineering, dry component assembly.
Certificazioni: EN 388:2016 (3131X)