Stampa…
Industrial gloves, Titan 385 / 388 / 391
Fornitore: MAPA
34388009 34388008 34388000
MAPA34388009EA 56.2 EUR
MAPA34388009 MAPA34388008 MAPA34388000
Industrial gloves, Titan 385 / 388 / 391
Guanti
These textile gloves either feature a ¾ or a full nitrile coating.
  • Freedom of movement: exceptionally comfortable textile support (no seams on the working surfaces)
  • Excellent resistance to oils and greases
  • Longer service life: quality of textile support and nitrile coating
  • Vented back models are available with or without knitted cuffs
Ideal for waste collection, forestry, metal shearing or cutting and the handling of thick, untrimmed sheet metal (not sharp-edged).
Certificazioni: EN 388:2016 (4111X); Cat. II