Stampa…
HPLC adapters for solvent supply and waste solvent
Fornitore: Merck
1.01111.0001 1.09996.0001 1.03833.0001 1.03830.0001 1.03831.0001
1.01111.0001EA 235 EUR
1.01111.0001 1.09996.0001 1.03833.0001 1.03830.0001 1.03831.0001
HPLC adapters for solvent supply and waste solvent
Ghiere Tappi di sicurezza e per rifiuti
Specially developed screw caps for direct instrument connection of Merck S 40 threaded bottles ensure safe handling of solvents in the lab and protect human health, the environment, as well as solvent quality.