Stampa…

2-Chloro-1,3,2-benzodioxaborole (β-bromocatecholborane) ≥96.0% (by titrimetric analysis), grado tecnico

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Chloro-1,3,2-benzodioxaborole, B-Chlorocatecholborane

C1669-25G C1669-5G
TCIAC1669-25GEA 496 EUR
TCIAC1669-25G TCIAC1669-5G
2-Chloro-1,3,2-benzodioxaborole (β-bromocatecholborane) ≥96.0% (by titrimetric analysis), grado tecnico
2-Chloro-1,3,2-benzodioxaborole (β-bromocatecholborane)
Formula: C₆H₄BClO₂
MDL Number: MFCD00043395
CAS Number: 55718-76-8
UN: 2925
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efde2065e64510fbad77c0ff89f3c4356d5b9eaf&vid=e565f4ebf3f458114186e72cc7487ad2b46c585c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (Argentmetric Titration) min. 96.0 %
Melting point 56.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR