Methyl-2,5-dibromobenzoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 2,5-dibromobenzoate
D2673-25G
D2673-5G
TCIAD2673-25GEA
117
EUR
TCIAD2673-25G
TCIAD2673-5G
Methyl-2,5-dibromobenzoate ≥98.0% (by GC)
Methyl-2,5-dibromobenzoate
Formula:
C₈H₆Br₂O₂ Peso molecolare: 293,94 g/mol |
MDL Number:
MFCD00144757 CAS Number: 57381-43-8 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...