Stampa…

Methyl-2-methyl-5-nitrobenzoate ≥98.0% (by GC)

Fornitore: TCI

M2220-25G M2220-5G
TCIAM2220-25GEA 87 EUR
TCIAM2220-25G TCIAM2220-5G
Methyl-2-methyl-5-nitrobenzoate ≥98.0% (by GC)
Methyl-2-methyl-5-nitrobenzoate
Formula: C₉H₉NO₄
MDL Number: MFCD00230865
CAS Number: 77324-87-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=516303c06ea0db21483f7676c3d9227e900ad8c8&vid=516303c06ea0db21483f7676c3d9227e900ad8c8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR