Stampa…

1-Phenyl-3-thiosemicarbazide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

P0234-1G P0234-25G
TCIAP0234-1GEA 16 EUR
TCIAP0234-1G TCIAP0234-25G
1-Phenyl-3-thiosemicarbazide ≥98.0% (by HPLC, titration analysis)
1-Phenyl-3-thiosemicarbazide
Formula: C₇H₉N₃S
MDL Number: MFCD00042739
CAS Number: 645-48-7
EINECS: 211-444-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f3415fda3c38989168eeba495aba498fbe64e84&vid=4f3415fda3c38989168eeba495aba498fbe64e84&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR