Stampa…

2-Bromo-4-nitroimidazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Bromo-5-nitro-1H-imidazole

B3503-1G B3503-5G
TCIAB3503-1GEA 35 EUR
TCIAB3503-1G TCIAB3503-5G
2-Bromo-4-nitroimidazole ≥98.0% (by GC)
2-Bromo-4-nitroimidazole
Formula: C₃H₂BrN₃O₂
Peso molecolare: 191,97 g/mol
MDL Number: MFCD09025308
CAS Number: 65902-59-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e933f8fdee0503d4a5debb3469bd51b60a1d653&vid=3e933f8fdee0503d4a5debb3469bd51b60a1d653&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 232.0 to 236.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR