4-Bromo-4'-iodobiphenyl ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4-Bromo-4'-iodo-1,1'-biphenyl, 1-Bromo-4-(4-iodophenyl)benzene
B3648-25G
B3648-5G
TCIAB3648-25GEA
88
EUR
TCIAB3648-25G
TCIAB3648-5G
4-Bromo-4'-iodobiphenyl ≥98.0% (by GC)
4-Bromo-4'-iodobiphenyl
Formula:
C₁₂H₈BrI Peso molecolare: 359,00 g/mol |
MDL Number:
MFCD17676204 CAS Number: 105946-82-5 UN: 3152 ADR: 9,II |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 175.0 to 179.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...