Stampa…

1-Fluoropyridiniumtrifluoromethanesulphonate ≥96.0% (by titrimetric analysis) fluorinating reagent

Fornitore: TCI EUROPE
Pericolo

Synonyms: N-Fluoropyridinium triflate, 1-Fluoropyridiniumtriflate, N-Fluoropyridinium trifluoromethanesulphonate

F0327-25G F0327-5G
TCIAF0327-25GEA 288 EUR
TCIAF0327-25G TCIAF0327-5G
1-Fluoropyridiniumtrifluoromethanesulphonate ≥96.0% (by titrimetric analysis) fluorinating reagent
1-Fluoropyridinium trifluoromethanesulphonate
Formula: C₆H₅F₄NO₃S
Peso molecolare: 247,17 g/mol
MDL Number: MFCD00013458
CAS Number: 107263-95-6
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32b8faf1c5275234373a5320df5e4ff577a6e215&vid=96c383170baa68644f5a7d3467b984787ab6af71&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Iodometric Titration) min. 96.0 %
Melting point 181.0 to 186.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR