1-Vinylimidazole ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₅H₆N₂ Punto di ebollizione: 80°C Flash Pt: 84°C |
MDL Number:
MFCD00005297 CAS Number: 1072-63-5 EINECS: 214-012-0 UN: 3267 ADR: 8,III |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...