Stampa…

1-Pyrenebutyric acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

P1213-1G P1213-5G
TCIAP1213-1GEA 86 EUR
TCIAP1213-1G TCIAP1213-5G
1-Pyrenebutyric acid ≥98.0% (by GC, titration analysis)
1-Pyrenebutyric acid
Formula: C₂₀H₁₆O₂
Peso molecolare: 288,35 g/mol
MDL Number: MFCD00004141
CAS Number: 3443-45-6
EINECS: 222-354-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a147f7eb827fb41aeb61d33e148290c6b0357fb&vid=6a147f7eb827fb41aeb61d33e148290c6b0357fb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 186.0 to 190.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR