Stampa…

Benzyl (S)-(+)-glycidyl ether ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-2-((Benzyloxy)methyl)oxirane

B2239-1G B2239-25G B2239-5G
TCIAB2239-1GEA 16 EUR
TCIAB2239-1G TCIAB2239-25G TCIAB2239-5G
Benzyl (S)-(+)-glycidyl ether ≥98.0% (by GC)
Benzyl (S)-(+)-glycidyl ether
Formula: C₁₀H₁₂O₂
Peso molecolare: 164,20 g/mol
MDL Number: MFCD00054428
CAS Number: 16495-13-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bde34d7980526ddbc36c0fbf26a403503f3ebf4d&vid=bde34d7980526ddbc36c0fbf26a403503f3ebf4d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +13.0 to +18.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR