Stampa…

(R)-(+)-Tetrahydro-2-furoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (R)-2-Tetrahydrofuroic acid

T1740-25G T1740-5G
TCIAT1740-25GEA 337 EUR
TCIAT1740-25G TCIAT1740-5G
(R)-(+)-Tetrahydro-2-furoic acid ≥98.0% (by GC, titration analysis)
(R)-(+)-Tetrahydro-2-furoic acid
Formula: C₅H₈O₃
Peso molecolare: 116,12 g/mol
Flash Pt: 139°C
MDL Number: MFCD00211271
CAS Number: 87392-05-0
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb4195957a0989f45b4de936180b48a29089dbc2&vid=fb4195957a0989f45b4de936180b48a29089dbc2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +18.0 to +20.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR