Stampa…
Butilacrilato ≥99.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
Pericolo
Synonyms: n-Butyl acrylate, Acrylic acid butyl ester
Stabilization: Stabilised with 4-methoxyphenol (MEHQ)
A0142-25ML A0142-500ML
TCIAA0142-25MLEA 16 EUR
TCIAA0142-25ML TCIAA0142-500ML
Butilacrilato ≥99.0% (by GC) stabilizzato
Butilacrilato
Formula: C₇H₁₂O₂
Peso molecolare: 128,17 g/mol
Flash Pt: 40°C
MDL Number: MFCD00008809
CAS Number: 141-32-2
EINECS: 205-480-7
UN: 2348
ADR: 3,III
Merck Index: 14,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=354f5004260ab2c1d7e5075cff45e6688db6d027&vid=354f5004260ab2c1d7e5075cff45e6688db6d027&prid=web&clid=SAPDAM
Visualizza i codici
Appearance Colorless clear liquid
Purity (GC) min. 99.0 %