Stampa…
D(+)-Glucosio 1 M aqueous solution, Sterile-Filtered
Fornitore: Thermo Fisher Scientific
Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
J60067.XE J60067.AK
J60067.XEEA 71.1 EUR
J60067.XE J60067.AK
D(+)-Glucosio 1 M aqueous solution, Sterile-Filtered
D(+)-Glucosio
CAS No.: 50-99-7 Molecular Formula: C6H12O6 Formula Weight: 180.16 Physical Form: Liquid MDL No.: MFCD00148912
Formula: C₆H₁₂O₆
Peso molecolare: 180.16 g/mol
Punto di fusione: 146
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM