Stampa…

5-Hydroxy-3,4-dimethoxybenzaldehyde ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Hydroxy-4,5-dimethoxybenzaldehyde, 3,4-Dimethoxy-5-hydroxybenzaldehyde

H1179-1G H1179-5G
TCIAH1179-1GEA 79 EUR
TCIAH1179-1G TCIAH1179-5G
5-Hydroxy-3,4-dimethoxybenzaldehyde ≥98.0% (by GC, titration analysis)
5-Hydroxy-3,4-dimethoxybenzaldehyde
Formula: C₉H₁₀O₄
Peso molecolare: 182,1733 g/mol
Punto di ebollizione: 180°C
MDL Number: MFCD00010442
CAS Number: 29865-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e282c74a11f5eed1cee60681de3bf403965624d&vid=8e282c74a11f5eed1cee60681de3bf403965624d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 64.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR