Stampa…

Ethyl arachidate ≥95.0% (by GC)

Fornitore: TCI

A0899-25G A0899-5G
TCIAA0899-25GEA 389 EUR
TCIAA0899-25G TCIAA0899-5G
Ethyl arachidate ≥95.0% (by GC)
Ethyl arachidate
Formula: C₂₂H₄₄O₂
Punto di ebollizione: 215°C
MDL Number: MFCD00056233
CAS Number: 18281-05-5
Merck Index: 14,00764
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05b3c0a2ebe80ecd93d64aaa82332c9eda33f189&vid=05b3c0a2ebe80ecd93d64aaa82332c9eda33f189&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 41.0 to 45.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR