6-Amino-1-naphthalenesulphonic acid ≥97.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-Amino-1-naphthalenesulphonic acid
A0347-25G
A0347-500G
TCIAA0347-25GEA
50
EUR
TCIAA0347-25G
TCIAA0347-500G
6-Amino-1-naphthalenesulphonic acid ≥97.0% (by HPLC, titration analysis)
6-Amino-1-naphthalenesulphonic acid
Formula:
C₁₀H₉NO₃S Peso molecolare: 223,2500 g/mol |
MDL Number:
MFCD00035724 CAS Number: 81-05-0 EINECS: 201-318-4 Merck Index: 14,06407 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light red powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...