7-Octenyl acetate ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₀H₁₈O₂ Punto di ebollizione: 104°C |
MDL Number:
MFCD01861278 CAS Number: 5048-35-1 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...