1-Chloro-2,5-diethoxybenzene ≥98.0% (by GC)
Fornitore: TCI EUROPE
C1576-25G
C1576-500G
TCIAC1576-25GEA
16
EUR
TCIAC1576-25G
TCIAC1576-500G
1-Chloro-2,5-diethoxybenzene ≥98.0% (by GC)
1-Chloro-2,5-diethoxybenzene
Formula:
C₁₀H₁₃ClO₂ Punto di ebollizione: 266°C Flash Pt: 130°C |
MDL Number:
MFCD00026765 CAS Number: 52196-74-4 EINECS: 257-728-9 |
Risultati del test di specificazione
Appearance | Colorless to Orange to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...