3-Chloromethyl-5-nitrosalicylaldehyde ≥97.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
3-(Chloromethyl)-2-hydroxy-5-nitrobenzaldehyde
C1305-25G
C1305-5G
TCIAC1305-25GEA
361
EUR
TCIAC1305-25G
TCIAC1305-5G
3-Chloromethyl-5-nitrosalicylaldehyde ≥97.0% (by HPLC, titration analysis)
3-Chloromethyl-5-nitrosalicylaldehyde
Formula:
C₈H₆ClNO₄ Peso molecolare: 215,5905 g/mol |
MDL Number:
MFCD00191331 CAS Number: 16644-30-7 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 90.0 to 93.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...