Stampa…

5-Chloro-3-nitrosalicylaldehyde ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

C2249-1G C2249-5G
TCIAC2249-1GEA 77 EUR
TCIAC2249-1G TCIAC2249-5G
5-Chloro-3-nitrosalicylaldehyde ≥98.0% (by GC, titration analysis)
5-Chloro-3-nitrosalicylaldehyde
Formula: C₇H₄ClNO₄
MDL Number: MFCD00462959
CAS Number: 16634-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=154aaf86d0615c815233572cbbec8cfba20e74da&vid=154aaf86d0615c815233572cbbec8cfba20e74da&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR