Stampa…

Ethyl isocyanatoacetate ≥97.0% (by GC)

Fornitore: TCI
Pericolo

I0386-25G I0386-5G
TCIAI0386-25GEA 173 EUR
TCIAI0386-25G TCIAI0386-5G
Ethyl isocyanatoacetate ≥97.0% (by GC)
Ethyl isocyanatoacetate
Formula: C₅H₇NO₃
Punto di ebollizione: 80°C
Flash Pt: 72°C
MDL Number: MFCD00002040
CAS Number: 2949-22-6
EINECS: 220-967-4
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d5271a7daae6220f220dae4eb2c357302aafd3&vid=90d5271a7daae6220f220dae4eb2c357302aafd3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Specific gravity (20/20) 1.1320 to 1.1360
Refractive index n20/D 1.4190 to 1.4230


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR