Stampa…

Tetraphenylphosphonium tetra-p-tolylborate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

T2603-25G T2603-5G
TCIAT2603-25GEA 256 EUR
TCIAT2603-25G TCIAT2603-5G
Tetraphenylphosphonium tetra-p-tolylborate ≥98.0% (by HPLC, titration analysis)
Tetraphenylphosphonium tetra-p-tolylborate
Formula: C₅₂H₄₈BP
MDL Number: MFCD08274957
CAS Number: 181259-35-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e86b40315ac686c1f6586fecf91611fc262c0f5a&vid=e86b40315ac686c1f6586fecf91611fc262c0f5a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR