1-(Methoxymethyl)benzotriazole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-(Methoxymethyl)-1H-benzotriazole
M1276-1G
M1276-5G
TCIAM1276-1GEA
73
EUR
TCIAM1276-1G
TCIAM1276-5G
1-(Methoxymethyl)benzotriazole ≥98.0% (by GC)
1-(Methoxymethyl)benzotriazole
Formula:
C₈H₉N₃O |
MDL Number:
MFCD00216630 CAS Number: 71878-80-3 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 39.0 to 43.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...