Stampa…

1-(Trimethylsilyl)benzotriazole ≥97.0% (by GC)

Fornitore: TCI
Pericolo

Synonyms: 1-(Trimethylsilyl)-1H-benzotriazole

T1752-25G T1752-5G
TCIAT1752-25GEA 166 EUR
TCIAT1752-25G TCIAT1752-5G
1-(Trimethylsilyl)benzotriazole ≥97.0% (by GC)
1-(Trimethylsilyl)benzotriazole
Formula: C₉H₁₃N₃Si
Flash Pt: –3°C
MDL Number: MFCD00216577
CAS Number: 43183-36-4
UN: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91a6536c9963ed55adeaa82ff67ca79b2ce56eb0&vid=91a6536c9963ed55adeaa82ff67ca79b2ce56eb0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR