Stampa…

1-Phenacylpyridinium bromide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: N-Phenacylpyridinium bromide, 1-(2-Oxo-2-phenylethyl)pyridin-1-ium bromide, N-(Benzoylmethyl)pyridinium bromide

P0797-25G P0797-5G
TCIAP0797-25GEA 59 EUR
TCIAP0797-25G TCIAP0797-5G
1-Phenacylpyridinium bromide ≥98.0% (by HPLC, titration analysis)
1-Phenacylpyridinium bromide
Formula: C₁₃H₁₂BrNO
Peso molecolare: 278.15 g/mol
MDL Number: MFCD00031995
CAS Number: 16883-69-5
EINECS: 240-919-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ee3525332f560bc0cf0500c91b969336b8ff1fb&vid=6ee3525332f560bc0cf0500c91b969336b8ff1fb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 198.0 to 203.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR