2-Hydroxyestradiol
Fornitore: Cayman Chemical
13019-1
13019-5
13019-10
13019-25
CAYM13019-1EA
96.2
EUR
CAYM13019-1
CAYM13019-5
CAYM13019-10
CAYM13019-25
2-Hydroxyestradiol
2-Hydroxyestradiol
Formula:
C₁₈H₂₄O₃ |
MDL Number:
MFCD00010490 CAS Number: 362-05-0 |