Stampa…

Estradiol ≥97.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: beta-Estradiol, 17β-Estradiol

E0025-1G E0025-25G E0025-5G
TCIAE0025-1GEA 33 EUR
TCIAE0025-1G TCIAE0025-25G TCIAE0025-5G
Estradiol ≥97.0% (by GC)
Estradiol
Formula: C₁₈H₂₄O₂
Peso molecolare: 272,39 g/mol
MDL Number: MFCD00003693
CAS Number: 50-28-2
EINECS: 200-023-8
Merck Index: 14,03703
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adab2dd7a69b43cc9fae1a739d0f84cad25a5fe5&vid=adab2dd7a69b43cc9fae1a739d0f84cad25a5fe5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Specific rotation [a]20/D +76.0 to +83.0 deg (C=1, Dioxane)
Melting point 176.0 to 180.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR