Stampa…

(S)-(-)-1-Boc-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (S)-1-Boc-3-Aminopyrrolidine, (S)-(-)-1-BOC-3-aminopyrrolidine, tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate

B2921-1G B2921-5G
TCIAB2921-1GEA 47 EUR
TCIAB2921-1G TCIAB2921-5G
(S)-(-)-1-Boc-3-aminopyrrolidine ≥98.0% (by GC, titration analysis)
(S)-(-)-1-Boc-3-aminopyrrolidine
Formula: C₉H₁₈N₂O₂
Peso molecolare: 186,25 g/mol
MDL Number: MFCD03419271
CAS Number: 147081-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d9d326286202e7ac16cfb49cdebbfeaff1a3931&vid=9d9d326286202e7ac16cfb49cdebbfeaff1a3931&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR