Stampa…
Filter tips, PIPETMAN® Diamond
Fornitore: GILSON
F171403 F171103 F171203 F171803 F171503 F171603 F171703 F171303 F172303 F172703 F172403 F172203 F172503 F172603 F172313 F172513 F172613 F172213 F172803
732-3402EA 145 EUR
732-3402 732-3399 732-3400 732-3406 732-3403 732-3404 732-3405 732-3401 GILSF172303 GILSF172703 GILSF172403 GILSF172203 GILSF172503 GILSF172603 GILSF172313 GILSF172513 GILSF172613 GILSF172213 732-3856
Filter tips, PIPETMAN® Diamond
Puntali per pipette
These filter tips made from 100% pure virgin PP are designed and manufactured in order to fit Gilson pipettes perfectly. They ensure optimal performance of PIPETMAN® pipettes across the full volume range. The high-density polyethylene filter provides an effective barrier against aerosols.
  • Smooth hydrophobic surface prevents liquid retention and ensures complete dispensing
  • Volume graduations make it easy to visually check the volume pipetted
  • No risk of contamination: Diamond tips are manufactured in a cleanroom
  • Gamma ray sterilised: Indicator dots used on Gilson's products undergo an irreversible colour change upon sterilisation
  • Guaranteed free of DNA, RNA, DNase, RNase, ATP, Pyrogens, dyes, additives and trace metals
  • Easy identification with colour-coded packaging
  • BLISTER refill: 45% less plastic than a regular rack box
TIPACKs are ready-to-use pipette tips in a rack, ready for autoclaving.
BLISTER refills are individually wrapped racks that can be used to quickly reload TIPACK boxes.
Informazioni per l'ordine: The empty boxes for the BLISTER refills need to be ordered separately.