Stampa…

2-(Hydroxymethyl)-12-crown-4 ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

H0982-1G H0982-5G
TCIAH0982-1GEA 587 EUR
TCIAH0982-1G TCIAH0982-5G
2-(Hydroxymethyl)-12-crown-4 ≥95.0% (by GC)
2-(Hydroxymethyl)-12-crown-4
Formula: C₉H₁₈O₅
Peso molecolare: 206,24 g/mol
MDL Number: MFCD00067524
CAS Number: 75507-26-5
EINECS: 278-224-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=297f1869cd13a8d815fd8baa8371ab655e6147ab&vid=297f1869cd13a8d815fd8baa8371ab655e6147ab&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR