Stampa…

3-Phenylbenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Biphenyl-3-carboxylic acid, 3-Biphenylcarboxylic acid, 1,1'-Biphenyl-3-carboxylic acid, [1,1'-Biphenyl]-3-carboxylic acid

B3850-5G B3850-1G
TCIAB3850-5GEA 89 EUR
TCIAB3850-5G TCIAB3850-1G
3-Phenylbenzoic acid ≥98.0% (by GC, titration analysis)
3-Phenylbenzoic acid
Formula: C₁₃H₁₀O₂
Peso molecolare: 198,22 g/mol
MDL Number: MFCD00045846
CAS Number: 716-76-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d6e0a8573d1b319c389a8bea31f863f2cb064f0&vid=5d6e0a8573d1b319c389a8bea31f863f2cb064f0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 164.0 to 168.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR