5-Bromo-2-fluoro-3-methylpyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Bromo-2-fluoro-3-picoline
B3775-5G
B3775-1G
TCIAB3775-5GEA
110
EUR
TCIAB3775-5G
TCIAB3775-1G
5-Bromo-2-fluoro-3-methylpyridine ≥98.0% (by GC)
5-Bromo-2-fluoro-3-methylpyridine
Formula:
C₆H₅BrFN Peso molecolare: 190,01 g/mol |
MDL Number:
MFCD03095091 CAS Number: 29312-98-9 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...