2-Aminonicotinonitrile ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Aminopyridine-3-carbonitrile, 2-Amino-3-cyanopyridine
A2443-5G
A2443-25G
TCIAA2443-5GEA
54
EUR
TCIAA2443-5G
TCIAA2443-25G
2-Aminonicotinonitrile ≥98.0% (by GC, titration analysis)
2-Aminonicotinonitrile
Formula:
C₆H₅N₃ Peso molecolare: 119,13 g/mol Punto di ebollizione: 268 °C (1013 hPa) Punto di fusione: 134 °C |
MDL Number:
MFCD00128867 CAS Number: 24517-64-4 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 133.0 to 137.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...